Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM49A
Entrez Gene ID
81553
Gene Name
family with sequence similarity 49 member A
Gene Aliases
-
Chromosome Location
Chr.2: 16549459 - 16665866 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030797.3 NP_110424.1 3 - 4 325 99
XM_005262630.3 XP_005262687.1 2 - 3 281 99
XM_006712108.3 XP_006712171.1 3 - 4 280 99
GenBank mRNA AK315065.1 - 2 - 3 220 99
AL136704.1 - 3 - 4 290 99
BC038971.1 - 3 - 4 276 99
CR533504.1 - 1 - 2 72 99
DA188616.1 - 3 - 4 325 99

View Full Product Details