The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030797.3 | NP_110424.1 | 8 - 9 | 885 | 65 |
XM_005262630.3 | XP_005262687.1 | 7 - 8 | 841 | 65 | |
XM_006712108.3 | XP_006712171.1 | 8 - 9 | 840 | 65 | |
GenBank mRNA | AK055334.1 | - | 8 - 9 | 853 | 65 |
AK091280.1 | - | 3 - 4 | 299 | 65 | |
AK315065.1 | - | 7 - 8 | 780 | 65 | |
AL136704.1 | - | 8 - 9 | 850 | 65 | |
BC038971.1 | - | 8 - 9 | 836 | 65 | |
CR533504.1 | - | 6 - 7 | 632 | 65 |