Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SIL1
Entrez Gene ID
64374
Gene Name
SIL1 nucleotide exchange factor
Gene Aliases
BAP, MSS, ULG5
Chromosome Location
Chr.5: 138946720 - 139198376 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037633.1 NP_001032722.1 2 - 3 192 107

View Full Product Details