The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037633.1 | NP_001032722.1 | 7 - 8 | 856 | 77 |
NM_022464.4 | NP_071909.1 | 6 - 7 | 768 | 77 | |
XM_011543570.2 | XP_011541872.1 | 8 - 9 | 893 | 77 | |
GenBank mRNA | AF547994.1 | - | 6 - 7 | 687 | 77 |
AJ299442.1 | - | 6 - 7 | 746 | 77 | |
AK074624.1 | - | 6 - 7 | 760 | 77 | |
AK075177.1 | - | 6 - 7 | 697 | 77 | |
AY358950.1 | - | 6 - 7 | 714 | 77 | |
BC011568.1 | - | 6 - 7 | 736 | 77 | |
KU178743.1 | - | 5 - 6 | 650 | 77 |