The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037633.1 | NP_001032722.1 | 9 - 10 | 1069 | 82 |
NM_022464.4 | NP_071909.1 | 8 - 9 | 981 | 82 | |
XM_011543570.2 | XP_011541872.1 | 10 - 11 | 1106 | 82 | |
GenBank mRNA | AF547994.1 | - | 8 - 9 | 900 | 82 |
AJ299442.1 | - | 8 - 9 | 959 | 82 | |
AK074624.1 | - | 8 - 9 | 973 | 82 | |
AK075177.1 | - | 8 - 9 | 910 | 82 | |
AY358950.1 | - | 8 - 9 | 927 | 82 | |
BC011568.1 | - | 8 - 9 | 949 | 82 | |
KU178743.1 | - | 7 - 8 | 863 | 82 | |
KU178744.1 | - | 6 - 7 | 671 | 82 |