The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270364.1 | NP_001257293.1 | 4 - 5 | 777 | 69 |
NM_012190.3 | NP_036322.2 | 4 - 5 | 879 | 69 | |
NR_072979.1 | 4 - 5 | 879 | 69 | ||
XM_006713481.2 | XP_006713544.1 | 4 - 5 | 618 | 69 | |
XM_011512355.1 | XP_011510657.1 | 4 - 5 | 611 | 69 | |
XM_017005613.1 | XP_016861102.1 | 4 - 5 | 1002 | 69 | |
XM_017005614.1 | XP_016861103.1 | 4 - 5 | 618 | 69 | |
GenBank mRNA | AB209299.1 | - | 4 - 5 | 686 | 69 |
AK222535.1 | - | 4 - 5 | 661 | 69 | |
AK222694.1 | - | 4 - 5 | 661 | 69 | |
AK294530.1 | - | 3 - 4 | 428 | 69 | |
AK307699.1 | - | 5 - 6 | 790 | 69 | |
AK308112.1 | - | 4 - 5 | 618 | 69 | |
BC027241.1 | - | 4 - 5 | 652 | 69 | |
KM111188.1 | - | 4 - 5 | 688 | 69 | |
KU178540.1 | - | 3 - 4 | 529 | 69 | |
KU178541.1 | - | 4 - 5 | 713 | 69 | |
KU178542.1 | - | 3 - 4 | 529 | 69 |