The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127173.1 | NP_001120645.1 | 3 - 4 | 521 | 64 |
NM_021189.3 | NP_067012.1 | 4 - 5 | 660 | 64 | |
GenBank mRNA | AF062733.2 | - | 4 - 5 | 617 | 64 |
AF363367.1 | - | 3 - 4 | 519 | 64 | |
AF529206.1 | - | 3 - 4 | 386 | 64 | |
AK289541.1 | - | 4 - 5 | 660 | 64 | |
AK308915.1 | - | 4 - 5 | 632 | 64 | |
AY046418.1 | - | 3 - 4 | 424 | 64 | |
AY358332.1 | - | 3 - 4 | 525 | 64 | |
BC033819.1 | - | 3 - 4 | 469 | 64 |