Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZC3H13
Entrez Gene ID
23091
Gene Name
zinc finger CCCH-type containing 13
Gene Aliases
KIAA0853
Chromosome Location
Chr.13: 45954469 - 46052785 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015070.4 NP_055885.3 16 - 17 4546 67
XM_005266301.3 XP_005266358.1 17 - 18 4924 67
XM_005266302.3 XP_005266359.1 17 - 18 4624 67
XM_005266303.3 XP_005266360.1 18 - 19 4949 67
XM_005266304.3 XP_005266361.1 17 - 18 4906 67
XM_005266305.3 XP_005266362.1 18 - 19 4942 67
XM_005266306.3 XP_005266363.1 17 - 18 4620 67
XM_005266307.3 XP_005266364.1 17 - 18 4921 67
XM_005266308.3 XP_005266365.1 16 - 17 4840 67
XM_005266309.3 XP_005266366.1 16 - 17 4540 67
XM_005266310.3 XP_005266367.1 16 - 17 4822 67
XM_005266311.3 XP_005266368.1 16 - 17 4837 67
XM_005266312.1 XP_005266369.1 16 - 17 4840 67
XM_017020475.1 XP_016875964.1 18 - 19 4712 67
XM_017020476.1 XP_016875965.1 18 - 19 4709 67
XM_017020477.1 XP_016875966.1 17 - 18 4865 67
XM_017020478.1 XP_016875967.1 17 - 18 4858 67
XM_017020479.1 XP_016875968.1 17 - 18 4628 67
XM_017020480.1 XP_016875969.1 16 - 17 4536 67
XM_017020481.1 XP_016875970.1 17 - 18 4862 67
XM_017020482.1 XP_016875971.1 16 - 17 4537 67
GenBank mRNA AB020660.1 - 5 - 6 2369 67
AK001815.1 - 2 - 3 231 67
AK074924.1 - 3 - 4 1597 67
AL136745.1 - 9 - 10 3582 67
AY283618.1 - 16 - 17 4544 67

View Full Product Details