Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DYSF
Entrez Gene ID
8291
Gene Name
dysferlin
Gene Aliases
FER1L1, LGMD2B, MMD1
Chromosome Location
Chr.2: 71453155 - 71686763 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130455.1 NP_001123927.1 40 - 41 4555 92
NM_001130976.1 NP_001124448.1 39 - 40 4745 92
NM_001130979.1 NP_001124451.1 41 - 42 4880 92
NM_001130980.1 NP_001124452.1 40 - 41 4838 92
NM_001130982.1 NP_001124454.1 41 - 42 4648 92
NM_001130985.1 NP_001124457.1 40 - 41 4606 92
NM_001130986.1 NP_001124458.1 39 - 40 4513 92
NM_003494.3 NP_003485.1 40 - 41 4787 92
GenBank mRNA AF075575.1 - 40 - 41 4784 92
AK074104.1 - 26 - 27 3057 92
DQ267935.1 - 40 - 41 4555 92
EU515155.1 - 41 - 42 4781 92
EU515156.1 - 39 - 40 4646 92
EU515158.1 - 40 - 41 4739 92
EU515161.1 - 41 - 42 4648 92
EU515162.1 - 39 - 40 4513 92
EU515164.1 - 40 - 41 4606 92

View Full Product Details