The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001294341.1 | NP_001281270.1 | 2 - 3 | 607 | 80 |
NM_032219.3 | NP_115595.2 | 2 - 3 | 607 | 80 | |
XM_005272299.4 | XP_005272356.1 | 2 - 3 | 634 | 80 | |
XM_006713918.1 | XP_006713981.1 | 2 - 3 | 261 | 80 | |
XM_011513568.1 | XP_011511870.1 | 2 - 3 | 1411 | 80 | |
XM_011513571.1 | XP_011511873.1 | 3 - 4 | 431 | 80 | |
XM_011513573.2 | XP_011511875.1 | 2 - 3 | 634 | 80 | |
XM_017008700.1 | XP_016864189.1 | 2 - 3 | 634 | 80 | |
GenBank mRNA | AK292010.1 | - | 2 - 3 | 505 | 80 |
AY358401.1 | - | 2 - 3 | 309 | 80 | |
BC030246.1 | - | 2 - 3 | 350 | 80 |