The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001018060.2 | NP_001018070.1 | 14 - 15 | 1498 | 109 |
NM_001146288.1 | NP_001139760.1 | 14 - 15 | 1583 | 109 | |
NM_144704.2 | NP_653305.1 | 14 - 15 | 1498 | 109 | |
NR_027464.1 | 13 - 14 | 1455 | 109 | ||
GenBank mRNA | AK055035.1 | - | 14 - 15 | 1482 | 109 |
AK094844.1 | - | 14 - 15 | 1487 | 109 | |
AK295581.1 | - | 13 - 14 | 1358 | 109 | |
AK316042.1 | - | 13 - 14 | 1351 | 109 | |
AY927594.1 | - | 5 - 6 | 109 | ||
BC032485.1 | - | 14 - 15 | 1465 | 109 | |
CR456342.1 | - | 13 - 14 | 1317 | 109 |