The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321053.1 | NP_001307982.1 | 2 - 3 | 424 | 71 |
NM_002727.3 | NP_002718.2 | 1 - 2 | 209 | 71 | |
XM_017016392.1 | XP_016871881.1 | 2 - 3 | 837 | 71 | |
GenBank mRNA | AK311873.1 | - | 1 - 2 | 121 | 71 |
BC015516.1 | - | 1 - 2 | 119 | 71 | |
BG548665.1 | - | 1 - 2 | 89 | 71 | |
BP335278.1 | - | 1 - 2 | 165 | 71 | |
BP363056.1 | - | 1 - 2 | 166 | 71 | |
CB110512.1 | - | 1 - 2 | 165 | 71 | |
J03223.1 | - | 1 - 2 | 110 | 71 | |
X17042.1 | - | 1 - 2 | 103 | 71 |