The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321041.1 | NP_001307970.1 | 3 - 4 | 565 | 85 |
NM_016321.2 | NP_057405.1 | 3 - 4 | 595 | 85 | |
NR_110261.1 | 3 - 4 | 595 | 85 | ||
GenBank mRNA | AF081497.1 | - | 3 - 4 | 566 | 85 |
AF193809.1 | - | 3 - 4 | 550 | 85 | |
AK290899.1 | - | 3 - 4 | 566 | 85 | |
AK298252.1 | - | 4 - 5 | 537 | 85 | |
AK301640.1 | - | 3 - 4 | 565 | 85 | |
AK310222.1 | - | 4 - 5 | 829 | 85 | |
AK313238.1 | - | 3 - 4 | 565 | 85 | |
AY257182.1 | - | 3 - 4 | 529 | 85 | |
BC030965.1 | - | 3 - 4 | 565 | 85 |