Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SUSD1
Entrez Gene ID
64420
Gene Name
sushi domain containing 1
Gene Aliases
-
Chromosome Location
Chr.9: 112040781 - 112175520 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282640.1 NP_001269569.1 8 - 9 1234 126
NM_001282643.1 NP_001269572.1 8 - 9 1234 126
NM_022486.4 NP_071931.2 8 - 9 1234 126
XM_017015038.1 XP_016870527.1 8 - 9 1457 126
GenBank mRNA AK025486.1 - 2 - 3 252 126
AK056704.1 - 8 - 9 1122 126
AK292868.1 - 8 - 9 1234 126
AL137432.1 - 4 - 5 641 126
AY358746.1 - 8 - 9 1213 126
BC052314.1 - 4 - 5 485 126
BC060770.1 - 8 - 9 1204 126

View Full Product Details