Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DOCK9
Entrez Gene ID
23348
Gene Name
dedicator of cytokinesis 9
Gene Aliases
ZIZ1, ZIZIMIN1
Chromosome Location
Chr.13: 98793487 - 99086406 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130048.1 NP_001123520.1 13 - 14 1679 73
NM_001130049.1 NP_001123521.1 13 - 14 1562 73
NM_001130050.1 NP_001123522.1 13 - 14 1679 73
NM_001318849.1 NP_001305778.1 13 - 14 1562 73
NM_015296.2 NP_056111.1 13 - 14 1562 73
XM_005254034.3 XP_005254091.1 13 - 14 1551 73
XM_005254035.3 XP_005254092.1 13 - 14 1551 73
XM_006719922.1 XP_006719985.1 13 - 14 1712 73
XM_006719924.2 XP_006719987.1 13 - 14 1551 73
XM_006719925.1 XP_006719988.1 13 - 14 1679 73
XM_006719931.3 XP_006719994.1 13 - 14 1507 73
XM_006719932.1 XP_006719995.1 13 - 14 1679 73
XM_006719936.1 XP_006719999.1 13 - 14 1712 73
XM_006719937.3 XP_006720000.1 13 - 14 1507 73
XM_017020503.1 XP_016875992.1 13 - 14 1552 73
XM_017020504.1 XP_016875993.1 13 - 14 1552 73
XM_017020505.1 XP_016875994.1 13 - 14 1552 73
XM_017020506.1 XP_016875995.1 13 - 14 1552 73
XM_017020507.1 XP_016875996.1 13 - 14 1552 73
XM_017020508.1 XP_016875997.1 13 - 14 1552 73
XM_017020509.1 XP_016875998.1 13 - 14 1679 73
XM_017020510.1 XP_016875999.1 13 - 14 1552 73
XM_017020511.1 XP_016876000.1 13 - 14 1552 73
XM_017020512.1 XP_016876001.1 13 - 14 1679 73
XM_017020513.1 XP_016876002.1 13 - 14 1552 73
XM_017020514.1 XP_016876003.1 13 - 14 1679 73
XM_017020515.1 XP_016876004.1 13 - 14 1679 73
XM_017020516.1 XP_016876005.1 13 - 14 1679 73
XM_017020517.1 XP_016876006.1 13 - 14 1651 73
XM_017020518.1 XP_016876007.1 13 - 14 1651 73
XM_017020519.1 XP_016876008.1 13 - 14 1679 73
XM_017020520.1 XP_016876009.1 13 - 14 1679 73
XM_017020521.1 XP_016876010.1 13 - 14 1712 73
GenBank mRNA AB028981.2 - 13 - 14 1585 73
AF527605.1 - 13 - 14 1562 73
AK122987.1 - 2 - 3 166 73
AK126492.1 - 13 - 14 1679 73
AK127329.1 - 13 - 14 1651 73
AK127508.1 - 10 - 11 1169 73
BC136274.1 - 13 - 14 1542 73
BC146814.1 - 13 - 14 1556 73
BC171753.1 - 13 - 14 1542 73
CR936820.1 - 13 - 14 1656 73

View Full Product Details