The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001035223.3 | NP_001030300.3 | 3 - 4 | 434 | 71 |
NM_001161616.2 | NP_001155088.2 | 3 - 4 | 434 | 71 | |
GenBank mRNA | AK293404.1 | - | 3 - 4 | 406 | 71 |
BC014426.1 | - | 3 - 4 | 422 | 71 | |
BC101756.1 | - | 3 - 4 | 420 | 71 | |
BC111958.1 | - | 3 - 4 | 420 | 71 | |
BC143530.1 | - | 3 - 4 | 420 | 71 | |
DB184397.1 | - | 3 - 4 | 436 | 71 |