The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286950.1 | NP_001273879.1 | 2 - 3 | 264 | 94 |
NM_001286954.1 | NP_001273883.1 | 2 - 3 | 264 | 94 | |
NM_144964.3 | NP_659401.2 | 2 - 3 | 264 | 94 | |
NR_104612.1 | 2 - 3 | 264 | 94 | ||
XM_011517735.2 | XP_011516037.1 | 2 - 3 | 286 | 94 | |
XM_011517736.2 | XP_011516038.1 | 2 - 3 | 291 | 94 | |
XM_011517742.1 | XP_011516044.1 | 2 - 3 | 304 | 94 | |
XM_011517743.2 | XP_011516045.1 | 2 - 3 | 304 | 94 | |
XM_017014313.1 | XP_016869802.1 | 2 - 3 | 304 | 94 | |
GenBank mRNA | AK056017.1 | - | 2 - 3 | 262 | 94 |
AK295733.1 | - | 2 - 3 | 254 | 94 | |
AK304516.1 | - | 2 - 3 | 259 | 94 | |
BC012175.1 | - | 2 - 3 | 250 | 94 | |
BC057774.1 | - | 2 - 3 | 251 | 94 | |
CR986760.1 | - | 2 - 3 | 251 | 94 |