The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317163.1 | NP_001304092.1 | 12 - 13 | 1869 | 133 |
NM_001317164.1 | NP_001304093.1 | 12 - 13 | 1540 | 133 | |
NM_001735.2 | NP_001726.2 | 12 - 13 | 1543 | 133 | |
XM_011518980.2 | XP_011517282.1 | 13 - 14 | 1912 | 133 | |
XM_017015102.1 | XP_016870591.1 | 14 - 15 | 2064 | 133 | |
XM_017015103.1 | XP_016870592.1 | 12 - 13 | 1697 | 133 | |
XM_017015104.1 | XP_016870593.1 | 14 - 15 | 2064 | 133 | |
GenBank mRNA | AB209031.1 | - | 12 - 13 | 1869 | 133 |
AK310774.1 | - | 12 - 13 | 1540 | 133 | |
AK310780.1 | - | 12 - 13 | 1542 | 133 | |
BC022299.1 | - | 12 - 13 | 1544 | 133 | |
BC113738.1 | - | 12 - 13 | 1538 | 133 | |
BC113740.1 | - | 12 - 13 | 1538 | 133 | |
M57729.1 | - | 12 - 13 | 1525 | 133 | |
M65134.1 | - | 2 - 3 | 280 | 133 |