The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317163.1 | NP_001304092.1 | 13 - 14 | 2071 | 116 |
NM_001317164.1 | NP_001304093.1 | 13 - 14 | 1742 | 116 | |
NM_001735.2 | NP_001726.2 | 13 - 14 | 1745 | 116 | |
XM_011518980.2 | XP_011517282.1 | 14 - 15 | 2114 | 116 | |
XM_017015102.1 | XP_016870591.1 | 15 - 16 | 2266 | 116 | |
XM_017015103.1 | XP_016870592.1 | 13 - 14 | 1899 | 116 | |
XM_017015104.1 | XP_016870593.1 | 15 - 16 | 2266 | 116 | |
GenBank mRNA | AB209031.1 | - | 13 - 14 | 2071 | 116 |
AK310774.1 | - | 13 - 14 | 1742 | 116 | |
AK310780.1 | - | 13 - 14 | 1744 | 116 | |
AV682721.1 | - | 1 - 2 | 205 | 116 | |
BC022299.1 | - | 13 - 14 | 1746 | 116 | |
BC113738.1 | - | 13 - 14 | 1740 | 116 | |
BC113740.1 | - | 13 - 14 | 1740 | 116 | |
M57729.1 | - | 13 - 14 | 1727 | 116 | |
M65134.1 | - | 3 - 4 | 482 | 116 |