The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317163.1 | NP_001304092.1 | 9 - 10 | 1358 | 144 |
NM_001317164.1 | NP_001304093.1 | 9 - 10 | 1029 | 144 | |
NM_001735.2 | NP_001726.2 | 9 - 10 | 1032 | 144 | |
XM_011518980.2 | XP_011517282.1 | 10 - 11 | 1401 | 144 | |
XM_017015102.1 | XP_016870591.1 | 11 - 12 | 1553 | 144 | |
XM_017015103.1 | XP_016870592.1 | 9 - 10 | 1186 | 144 | |
XM_017015104.1 | XP_016870593.1 | 11 - 12 | 1553 | 144 | |
GenBank mRNA | AB209031.1 | - | 9 - 10 | 1358 | 144 |
AK310774.1 | - | 9 - 10 | 1029 | 144 | |
AK310780.1 | - | 9 - 10 | 1031 | 144 | |
BC022299.1 | - | 9 - 10 | 1033 | 144 | |
BC113738.1 | - | 9 - 10 | 1027 | 144 | |
BC113740.1 | - | 9 - 10 | 1027 | 144 | |
M57729.1 | - | 9 - 10 | 1014 | 144 |