Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PFAS
Entrez Gene ID
5198
Gene Name
phosphoribosylformylglycinamidine synthase
Gene Aliases
FGAMS, FGAR-AT, FGARAT, PURL
Chromosome Location
Chr.17: 8249255 - 8270491 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012393.2 NP_036525.1 27 - 28 3847 90
XM_017024760.1 XP_016880249.1 26 - 27 3649 90
GenBank mRNA AB002359.1 - 27 - 28 3806 90
AK292402.1 - 27 - 28 3841 90
AK292804.1 - 27 - 28 3847 90
AK295895.1 - 25 - 26 3765 90
BC006522.2 - 9 - 10 1374 90
BC063538.1 - 13 - 14 1938 90
BC146768.1 - 27 - 28 3806 90
BC167158.1 - 27 - 28 3806 90

View Full Product Details