The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001143818.1 | NP_001137290.1 | 5 - 6 | 749 | 134 |
NM_002575.2 | NP_002566.1 | 4 - 5 | 491 | 134 | |
GenBank mRNA | A21238.1 | - | 4 - 5 | 464 | 134 |
A32415.1 | - | 4 - 5 | 457 | 134 | |
AK313159.1 | - | 4 - 5 | 497 | 134 | |
BC012609.1 | - | 4 - 5 | 495 | 134 | |
J02685.1 | - | 4 - 5 | 471 | 134 | |
J03603.1 | - | 4 - 5 | 464 | 134 | |
M18082.1 | - | 3 - 4 | 447 | 134 | |
Y00630.1 | - | 4 - 5 | 488 | 134 |