The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001267040.1 | NP_001253969.1 | 9 - 10 | 2605 | 81 |
NM_004973.3 | NP_004964.2 | 9 - 10 | 3105 | 81 | |
XM_005249089.3 | XP_005249146.1 | 8 - 9 | 2443 | 81 | |
XM_011514579.2 | XP_011512881.1 | 11 - 12 | 3294 | 81 | |
XM_011514584.2 | XP_011512886.1 | 7 - 8 | 2359 | 81 | |
XM_017010833.1 | XP_016866322.1 | 11 - 12 | 3249 | 81 | |
XM_017010834.1 | XP_016866323.1 | 8 - 9 | 2963 | 81 | |
XM_017010835.1 | XP_016866324.1 | 7 - 8 | 2338 | 81 | |
GenBank mRNA | AK292861.1 | - | 9 - 10 | 2951 | 81 |
AK299349.1 | - | 8 - 9 | 2519 | 81 | |
AK303610.1 | - | 9 - 10 | 2548 | 81 | |
BC046246.1 | - | 9 - 10 | 2822 | 81 | |
U57592.1 | - | 9 - 10 | 2784 | 81 |