The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001220488.1 | NP_001207417.1 | 6 - 7 | 1039 | 76 |
NM_001220489.1 | NP_001207418.1 | 4 - 5 | 816 | 76 | |
NM_001220490.1 | NP_001207419.1 | 4 - 5 | 724 | 76 | |
NM_001257.4 | NP_001248.1 | 5 - 6 | 933 | 76 | |
XM_011522804.2 | XP_011521106.1 | 4 - 5 | 540 | 76 | |
XM_017022848.1 | XP_016878337.1 | 6 - 7 | 956 | 76 | |
GenBank mRNA | AK295886.1 | - | 4 - 5 | 554 | 76 |
AK298612.1 | - | 4 - 5 | 642 | 76 | |
AK304733.1 | - | 6 - 7 | 794 | 76 | |
AK307862.1 | - | 6 - 7 | 730 | 76 | |
BC028624.2 | - | 5 - 6 | 725 | 76 | |
BC030653.2 | - | 5 - 6 | 725 | 76 | |
BU162665.1 | - | 1 - 2 | 66 | 76 | |
L34058.1 | - | 5 - 6 | 1078 | 76 | |
U59288.1 | - | 5 - 6 | 850 | 76 |