Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NOP16
Entrez Gene ID
51491
Gene Name
NOP16 nucleolar protein
Gene Aliases
HSPC111, HSPC185
Chromosome Location
Chr.5: 176383937 - 176388927 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256539.3 NP_001243468.2 1 - 2 491 77
NM_001256540.3 NP_001243469.2 1 - 2 491 77
NM_001291305.2 NP_001278234.1 1 - 2 491 77
NM_001291307.2 NP_001278236.1 1 - 2 491 77
NM_001291308.2 NP_001278237.1 1 - 2 491 77
NM_016391.7 NP_057475.2 1 - 2 491 77
NR_134318.1 1 - 2 491 77
XM_011534567.1 XP_011532869.1 1 - 2 170 77
GenBank mRNA AF151019.1 - 1 - 2 77
AF161460.1 - 1 - 2 77
AK300883.1 - 1 - 2 170 77
AK303281.1 - 1 - 2 162 77
AY561704.1 - 1 - 2 103 77
BC019331.1 - 1 - 2 326 77
BC032424.2 - 1 - 2 134 77
BC040106.1 - 1 - 2 151 77
BM826387.1 - 1 - 2 150 77
BU594763.1 - 1 - 2 165 77
CB122385.1 - 1 - 2 151 77
CR457169.1 - 1 - 2 103 77
HY064071.1 - 1 - 2 174 77

View Full Product Details