Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ADAMTS19
Entrez Gene ID
171019
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 19
Gene Aliases
-
Chromosome Location
Chr.5: 129460281 - 129738683 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011543246.2 XP_011541548.1 13 - 14 1685 109
XM_011543247.2 XP_011541549.1 13 - 14 1709 109
XM_011543248.2 XP_011541550.1 13 - 14 1829 109
XM_011543249.2 XP_011541551.1 8 - 9 1988 109
XM_017009174.1 XP_016864663.1 12 - 13 2444 109
XM_017009175.1 XP_016864664.1 9 - 10 2741 109
XM_017009176.1 XP_016864665.1 14 - 15 1857 109
GenBank mRNA AJ311904.1 - 14 - 15 2285 109
AK122622.1 - 4 - 5 531 109
AK128729.1 - 12 - 13 1500 109

View Full Product Details