The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318480.1 | NP_001305409.1 | 6 - 7 | 895 | 83 |
NM_018986.4 | NP_061859.3 | 6 - 7 | 812 | 83 | |
XM_006713889.2 | XP_006713952.1 | 3 - 4 | 230 | 83 | |
XM_011513488.1 | XP_011511790.1 | 6 - 7 | 789 | 83 | |
XM_017008300.1 | XP_016863789.1 | 6 - 7 | 921 | 83 | |
XM_017008301.1 | XP_016863790.1 | 6 - 7 | 685 | 83 | |
XM_017008302.1 | XP_016863791.1 | 3 - 4 | 230 | 83 | |
GenBank mRNA | AK000363.1 | - | 2 - 3 | 200 | 83 |
AK074402.1 | - | 6 - 7 | 689 | 83 | |
AK126165.1 | - | 6 - 7 | 768 | 83 | |
AK222865.1 | - | 2 - 3 | 200 | 83 |