The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001242929.1 | NP_001229858.1 | 1 - 2 | 220 | 105 |
NR_040252.1 | 1 - 2 | 220 | 105 | ||
XM_005255119.3 | XP_005255176.1 | 1 - 2 | 190 | 105 | |
XM_005255121.3 | XP_005255178.1 | 1 - 2 | 92 | 105 | |
XM_011522373.1 | XP_011520675.1 | 1 - 2 | 103 | 105 | |
GenBank mRNA | AK123371.1 | - | 1 - 2 | 91 | 104 |
DR004376.1 | - | 1 - 2 | 226 | 104 |