Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PDIA2
Entrez Gene ID
64714
Gene Name
protein disulfide isomerase family A member 2
Gene Aliases
PDA2, PDI, PDIP, PDIR
Chromosome Location
Chr.16: 283118 - 287209 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006849.2 NP_006840.2 5 - 6 847 53
GenBank mRNA AK075485.1 - 6 - 7 764 53
AK124931.1 - 4 - 5 866 53
AK295383.1 - 5 - 6 739 53
AK308516.1 - 5 - 6 727 53
BC000537.2 - 5 - 6 781 53
BC075029.2 - 5 - 6 779 53
U19948.1 - 5 - 6 780 53

View Full Product Details