The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001176.3 | NP_001167.2 | 3 - 4 | 413 | 94 |
GenBank mRNA | AB593111.1 | - | 3 - 4 | 572 | 94 |
AF498928.1 | - | 3 - 4 | 338 | 94 | |
BC047699.1 | - | 3 - 4 | 414 | 94 |