Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NR3C1
Entrez Gene ID
2908
Gene Name
nuclear receptor subfamily 3 group C member 1
Gene Aliases
GCCR, GCR, GCRST, GR, GRL
Chromosome Location
Chr.5: 143277931 - 143435512 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000176.2 NP_000167.1 6 - 7 2383 66
NM_001018074.1 NP_001018084.1 6 - 7 2213 66
NM_001018075.1 NP_001018085.1 6 - 7 2116 66
NM_001018076.1 NP_001018086.1 6 - 7 2009 66
NM_001018077.1 NP_001018087.1 6 - 7 2885 66
NM_001020825.1 NP_001018661.1 6 - 7 2383 66
NM_001024094.1 NP_001019265.1 6 - 7 2386 66
NM_001204258.1 NP_001191187.1 6 - 7 2383 66
NM_001204259.1 NP_001191188.1 6 - 7 2383 66
NM_001204260.1 NP_001191189.1 6 - 7 2383 66
NM_001204261.1 NP_001191190.1 6 - 7 2383 66
NM_001204262.1 NP_001191191.1 6 - 7 2383 66
NM_001204263.1 NP_001191192.1 6 - 7 2383 66
NM_001204264.1 NP_001191193.1 6 - 7 2383 66
NM_001204265.1 NP_001191194.1 6 - 7 2383 66
XM_005268419.3 XP_005268476.1 6 - 7 2116 66
XM_005268420.4 XP_005268477.1 6 - 7 2297 66
XM_005268422.3 XP_005268479.1 6 - 7 2178 66
XM_005268423.3 XP_005268480.1 6 - 7 2167 66
XM_011537637.2 XP_011535939.1 5 - 6 2818 66
XM_017009397.1 XP_016864886.1 6 - 7 2009 66
XM_017009398.1 XP_016864887.1 6 - 7 2308 66
GenBank mRNA AB307716.1 - 5 - 6 1891 66
AK223594.1 - 6 - 7 2237 66
AK302050.1 - 5 - 6 802 66
AK314789.1 - 6 - 7 2013 66
AM183262.1 - 6 - 7 1826 66
BC015610.2 - 6 - 7 2331 66
BX640610.1 - 6 - 7 2074 66
BX648553.1 - 7 - 8 2178 66
HQ450643.1 - 5 - 6 1891 66
HQ450644.1 - 5 - 6 1891 66
HQ692822.1 - 5 - 6 1891 66
JN797823.1 - 5 - 6 1890 66
JN797824.1 - 5 - 6 1891 66
U01351.1 - 6 - 7 2387 66
X03225.1 - 6 - 7 2023 66
X03348.1 - 6 - 7 2023 66

View Full Product Details