The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018079.4 | NP_060549.4 | 15 - 16 | 2041 | 83 |
GenBank mRNA | AK001241.1 | - | 9 - 10 | 938 | 83 |
AK301122.1 | - | 7 - 8 | 740 | 83 | |
AK316222.1 | - | 7 - 8 | 721 | 83 | |
BC032538.1 | - | 9 - 10 | 1016 | 83 |