The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018079.4 | NP_060549.4 | 5 - 6 | 887 | 103 |
XM_011532946.1 | XP_011531248.1 | 5 - 6 | 888 | 103 | |
XM_017004417.1 | XP_016859906.1 | 5 - 6 | 888 | 103 | |
GenBank mRNA | AK056536.1 | - | 5 - 6 | 857 | 103 |