The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004422.2 | NP_004413.1 | 7 - 8 | 1098 | 109 |
XM_005256502.1 | XP_005256559.1 | 7 - 8 | 1090 | 109 | |
XM_017024298.1 | XP_016879787.1 | 4 - 5 | 444 | 109 | |
GenBank mRNA | AF006012.1 | - | 7 - 8 | 822 | 109 |
AK026019.1 | - | 6 - 7 | 624 | 109 | |
AK297099.1 | - | 7 - 8 | 854 | 109 | |
AK297285.1 | - | 7 - 8 | 831 | 109 | |
AK304229.1 | - | 7 - 8 | 865 | 109 | |
AK312829.1 | - | 7 - 8 | 1099 | 109 | |
BC014844.1 | - | 7 - 8 | 911 | 109 | |
BT009822.1 | - | 7 - 8 | 816 | 109 |