Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DYNC2LI1
Entrez Gene ID
51626
Gene Name
dynein cytoplasmic 2 light intermediate chain 1
Gene Aliases
CGI-60, D2LIC, LIC3
Chromosome Location
Chr.2: 43774039 - 43828492 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193464.1 NP_001180393.1 1 - 2 110 58
NM_015522.3 NP_056337.1 1 - 2 110 58
NM_016008.3 NP_057092.2 1 - 2 110 58
XM_005264364.4 XP_005264421.1 1 - 2 87 58
XM_005264365.4 XP_005264422.1 1 - 2 87 58
GenBank mRNA AF151818.1 - 1 - 2 71 58
AK074206.1 - 1 - 2 100 58
AK223432.1 - 1 - 2 110 58
AK301489.1 - 1 - 2 100 58
AY083346.1 - 1 - 2 70 58
BC006969.1 - 1 - 2 69 58
BC016324.1 - 1 - 2 104 58
BC040558.1 - 1 - 2 85 58
BC058823.1 - 1 - 2 77 58
BG716986.1 - 1 - 2 115 58
BX493698.1 - 1 - 2 113 58

View Full Product Details