Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NXF4
Entrez Gene ID
55999
Gene Name
nuclear RNA export factor 4 pseudogene
Gene Aliases
-
Chromosome Location
Chr.X: 102549965 - 102571693 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_002216.1 13 - 14 1532 107
GenBank mRNA AK124700.1 - 13 - 14 1532 107


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream thymosin beta 15a TMSB15A TMSB15, TMSB15B, TMSL8, TMSNB, Tb15, TbNB -49.7kb Anti-sense Chr. - 102513682 - 102516771 on Build GRCh38
Downstream armadillo repeat containing, X-linked 5 ARMCX5 GASP5 32.4kb Sense Chr. - 102599168 - 102604159 on Build GRCh38

View Full Product Details