The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001168478.1 | NP_001161950.1 | 1 - 2 | 348 | 151 |
NM_001168479.1 | NP_001161951.1 | 3 - 4 | 464 | 151 | |
NM_001168480.1 | NP_001161952.1 | 3 - 4 | 458 | 151 | |
GenBank mRNA | AK096692.1 | - | 3 - 4 | 458 | 151 |
AL537480.3 | - | 1 - 2 | 268 | 151 | |
BC050678.1 | - | 1 - 2 | 309 | 151 | |
BI765968.1 | - | 3 - 4 | 416 | 151 |