Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
STRN
Entrez Gene ID
6801
Gene Name
striatin
Gene Aliases
PPP2R6A, SG2NA
Chromosome Location
Chr.2: 36837698 - 36966607 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003162.3 NP_003153.2 14 - 15 1847 78
XM_005264519.4 XP_005264576.1 13 - 14 1871 78
XM_011533073.2 XP_011531375.1 15 - 16 2069 78
GenBank mRNA AJ223814.1 - 14 - 15 78
AK026934.1 - 11 - 12 1418 78
BC106878.2 - 15 - 16 1764 78
BC106879.2 - 14 - 15 1699 78

View Full Product Details