The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004259.6 | NP_004250.4 | 10 - 11 | 1707 | 118 |
XM_005257818.3 | XP_005257875.1 | 10 - 11 | 1720 | 118 | |
XM_005257822.3 | XP_005257879.1 | 3 - 4 | 1078 | 118 | |
XM_005257823.3 | XP_005257880.1 | 10 - 11 | 1719 | 118 | |
XM_006722186.2 | XP_006722249.1 | 3 - 4 | 405 | 118 | |
XM_011525482.2 | XP_011523784.1 | 9 - 10 | 2187 | 118 | |
XM_011525484.1 | XP_011523786.1 | 4 - 5 | 662 | 118 | |
XM_011525485.2 | XP_011523787.1 | 4 - 5 | 601 | 118 | |
XM_011525486.2 | XP_011523788.1 | 3 - 4 | 588 | 118 | |
XM_017025343.1 | XP_016880832.1 | 9 - 10 | 2187 | 118 | |
GenBank mRNA | AB042824.1 | - | 10 - 11 | 1707 | 118 |
AF193041.1 | - | 3 - 4 | 1083 | 118 | |
AK304168.1 | - | 10 - 11 | 1699 | 118 | |
BC000570.2 | - | 4 - 5 | 472 | 118 | |
BC063440.1 | - | 10 - 11 | 1579 | 118 | |
EF560729.1 | - | 10 - 11 | 1672 | 118 |