The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001251902.1 | NP_001238831.1 | 2 - 3 | 398 | 91 |
NM_003985.4 | NP_003976.2 | 2 - 3 | 398 | 91 | |
XM_011524045.2 | XP_011522347.1 | 2 - 3 | 446 | 91 | |
XM_011524046.2 | XP_011522348.1 | 2 - 3 | 535 | 91 | |
XM_011524047.2 | XP_011522349.1 | 2 - 3 | 535 | 91 | |
GenBank mRNA | AK225403.1 | - | 2 - 3 | 281 | 91 |
AK310684.1 | - | 2 - 3 | 281 | 91 | |
DA089305.1 | - | 2 - 3 | 535 | 91 |