Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WSCD1
Entrez Gene ID
23302
Gene Name
WSC domain containing 1
Gene Aliases
-
Chromosome Location
Chr.17: 6069106 - 6124427 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015253.1 NP_056068.1 2 - 3 754 117
XM_005256572.1 XP_005256629.1 2 - 3 842 117
XM_005256573.1 XP_005256630.1 2 - 3 817 117
XM_011523777.1 XP_011522079.1 2 - 3 842 117
GenBank mRNA AB011095.1 - 1 - 2 106 117
AK289603.1 - 2 - 3 813 117
AL833730.1 - 2 - 3 754 117
BC009975.2 - 2 - 3 779 117

View Full Product Details