The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004104.4 | NP_004095.4 | 19 - 20 | 3163 | 79 |
XM_011523538.2 | XP_011521840.1 | 19 - 20 | 3231 | 79 | |
GenBank mRNA | AB209988.1 | - | 19 - 20 | 3158 | 79 |
AY451392.1 | - | 18 - 19 | 3046 | 79 | |
BC063242.1 | - | 19 - 20 | 3163 | 79 | |
U26644.1 | - | 18 - 19 | 3040 | 79 | |
U29344.1 | - | 19 - 20 | 3169 | 79 |