Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FASN
Entrez Gene ID
2194
Gene Name
fatty acid synthase
Gene Aliases
FAS, OA-519, SDR27X1
Chromosome Location
Chr.17: 82078338 - 82098303 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004104.4 NP_004095.4 24 - 25 4401 75
XM_011523538.2 XP_011521840.1 24 - 25 4469 75
GenBank mRNA AB209988.1 - 24 - 25 4396 75
AY451392.1 - 23 - 24 4284 75
BC014631.2 - 4 - 5 1003 75
BC063242.1 - 24 - 25 4401 75
U26644.1 - 23 - 24 4275 75
U29344.1 - 24 - 25 4404 75

View Full Product Details