The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004104.4 | NP_004095.4 | 28 - 29 | 5039 | 83 |
XM_011523538.2 | XP_011521840.1 | 28 - 29 | 5107 | 83 | |
GenBank mRNA | AB209988.1 | - | 28 - 29 | 5034 | 83 |
AY451392.1 | - | 27 - 28 | 4922 | 83 | |
BC007267.1 | - | 2 - 3 | 181 | 83 | |
BC014631.2 | - | 8 - 9 | 1641 | 83 | |
BC063242.1 | - | 28 - 29 | 5039 | 83 | |
U26644.1 | - | 27 - 28 | 4907 | 83 | |
U29344.1 | - | 28 - 29 | 5039 | 83 |