The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004104.4 | NP_004095.4 | 34 - 35 | 6036 | 53 |
XM_011523538.2 | XP_011521840.1 | 34 - 35 | 6104 | 53 | |
GenBank mRNA | AB209988.1 | - | 34 - 35 | 6031 | 53 |
AY451392.1 | - | 33 - 34 | 5919 | 53 | |
BC007267.1 | - | 8 - 9 | 1178 | 53 | |
BC007909.1 | - | 3 - 4 | 451 | 53 | |
BC063242.1 | - | 34 - 35 | 6036 | 53 | |
U29344.1 | - | 34 - 35 | 6036 | 53 |