The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004104.4 | NP_004095.4 | 39 - 40 | 6950 | 66 |
XM_011523538.2 | XP_011521840.1 | 39 - 40 | 7018 | 66 | |
GenBank mRNA | AB209988.1 | - | 39 - 40 | 6945 | 66 |
AY451392.1 | - | 38 - 39 | 6833 | 66 | |
BC007267.1 | - | 13 - 14 | 2092 | 66 | |
BC007909.1 | - | 8 - 9 | 1365 | 66 | |
BC014631.2 | - | 15 - 16 | 2939 | 66 | |
BC014634.2 | - | 2 - 3 | 361 | 66 | |
BC063242.1 | - | 39 - 40 | 6950 | 66 | |
S80437.1 | - | 4 - 5 | 704 | 66 | |
U29344.1 | - | 39 - 40 | 6950 | 66 |