Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TTC38
Entrez Gene ID
55020
Gene Name
tetratricopeptide repeat domain 38
Gene Aliases
LL22NC03-5H6.5
Chromosome Location
Chr.22: 46267964 - 46294008 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011530259.1 XP_011528561.1 2 - 3 170 128

View Full Product Details