Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PCM1
Entrez Gene ID
5108
Gene Name
pericentriolar material 1
Gene Aliases
PTC4, RET/PCM-1
Chromosome Location
Chr.8: 17922763 - 18029948 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001315507.1 NP_001302436.1 6 - 7 1210 117
NM_001315508.1 NP_001302437.1 6 - 7 1210 117
NM_006197.3 NP_006188.3 6 - 7 1210 117
XM_006716349.3 XP_006716412.1 6 - 7 1283 117
XM_017013479.1 XP_016868968.1 8 - 9 1587 117
XM_017013480.1 XP_016868969.1 6 - 7 1283 117
XM_017013481.1 XP_016868970.1 8 - 9 1586 117
XM_017013482.1 XP_016868971.1 6 - 7 1304 117
XM_017013483.1 XP_016868972.1 8 - 9 1585 117
XM_017013484.1 XP_016868973.1 5 - 6 1011 117
XM_017013485.1 XP_016868974.1 8 - 9 1586 117
XM_017013491.1 XP_016868980.1 6 - 7 1283 117
XM_017013492.1 XP_016868981.1 6 - 7 1283 117
XM_017013493.1 XP_016868982.1 8 - 9 1586 117
XM_017013494.1 XP_016868983.1 6 - 7 1283 117
XM_017013495.1 XP_016868984.1 8 - 9 1423 117
XM_017013496.1 XP_016868985.1 5 - 6 1006 117
XM_017013497.1 XP_016868986.1 6 - 7 1283 117
XM_017013498.1 XP_016868987.1 6 - 7 1283 117
XM_017013499.1 XP_016868988.1 8 - 9 1423 117
XM_017013500.1 XP_016868989.1 6 - 7 1283 117
XM_017013501.1 XP_016868990.1 8 - 9 1585 117
XM_017013502.1 XP_016868991.1 5 - 6 1006 117
XM_017013504.1 XP_016868993.1 6 - 7 1304 117
XM_017013505.1 XP_016868994.1 8 - 9 1423 117
XM_017013506.1 XP_016868995.1 6 - 7 1283 117
XM_017013507.1 XP_016868996.1 8 - 9 1587 117
XM_017013508.1 XP_016868997.1 7 - 8 1227 117
GenBank mRNA AX785390.1 - 4 - 5 788 117
BC027477.1 - 5 - 6 989 117
BC133052.1 - 5 - 6 863 117
BC140946.1 - 6 - 7 1039 117
L27841.1 - 6 - 7 1197 117

View Full Product Details