Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SPTA1
Entrez Gene ID
6708
Gene Name
spectrin alpha, erythrocytic 1
Gene Aliases
EL2, HPP, HS3, SPH3, SPTA
Chromosome Location
Chr.1: 158610498 - 158686716 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003126.2 NP_003117.2 46 - 47 6746 79
XM_011509916.2 XP_011508218.1 47 - 48 6624 79
GenBank mRNA M61877.1 - 46 - 47 6730 79

View Full Product Details