Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM160B2
Entrez Gene ID
64760
Gene Name
family with sequence similarity 160 member B2
Gene Aliases
RAI16
Chromosome Location
Chr.8: 22089150 - 22104380 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022749.5 NP_073586.5 1 - 2 96 135
XM_005273602.3 XP_005273659.1 1 - 2 96 135
XM_006716381.2 XP_006716444.1 1 - 2 96 135
XM_017013732.1 XP_016869221.1 1 - 2 96 135
GenBank mRNA AK126408.1 - 1 - 2 96 135
BC052237.1 - 1 - 2 93 135

View Full Product Details